Läs mer
Desmethyl Dehydro Lercanidipine (>90%), TRC
Produktkod. 30353670
Change view
Kvantitet:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30353670 5 mg
30367638 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30353670 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2909545MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 3,5-Pyridinedicarboxylic acid, 2,6-dimethyl-4-(3-nitrophenyl)-, 3-[2-[(3,3-diphenylpropyl)methylamino]-1,1-dimethylethyl] ester
CAS 1119226-97-9
Molekylformel C35 H37 N3 O6
Kvantitet 5 mg
Synonym 3-[2-[(3,3-Diphenylpropyl)methylamino]-1,1-dimethylethyl] hydrogen 2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylate, Lercanidipine Impurity B
InChI formel InChI=1S/C35H37N3O6/c1-23-30(33(39)40)32(27-17-12-18-28(21-27)38(42)43)31(24(2)36-23)34(41)44-35(3,4)22-37(5)20-19-29(25-13-8-6-9-14-25)26-15-10-7-11-16-26/h6-18,21,29H,19-20,22H2,1-5H3,(H,39,40)
LEDER CN(CCC(c1ccccc1)c2ccccc2)CC(C)(C)OC(=O)c3c(C)nc(C)c(C(=O)O)c3c4cccc(c4)[N+](=O)[O-]
IUPAC-namn 5-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl]oxycarbonyl-2,6-dimethyl-4-(3-nitrophenyl)pyridine-3-carboxylic acid
Molekylvikt (g/mol) 595.68
Formel vikt 595.2682