Läs mer
Desmethyl Erlotinib Acetate, TRC
Produktkod. 30137057
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137057 10 mg
30009573 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137057 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29202010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 183320-15-2
Molekylformel C23 H23 N3 O5
Molekylvikt (g/mol) 421.45
IUPAC-namn 2-[4-(3-ethynylanilino)-7-(2-methoxyethoxy)quinazolin-6-yl]oxyethyl acetate
LEDER COCCOc1cc2ncnc(Nc3cccc(c3)C#C)c2cc1OCCOC(=O)C
Specifikationer
Kemiskt namn eller material Desmethyl Erlotinib Acetate
Analyt- eller komponentnamn Desmethyl Erlotinib Acetate
Frakt skick Room Temperature
InChI formel InChI=1S/C23H23N3O5/c1-4-17-6-5-7-18(12-17)26-23-19-13-21(31-11-10-29-16(2)27)22(30-9-8-28-3)14-20(19)24-15-25-23/h1,5-7,12-15H,8-11H2,2-3H3,(H,24,25,26)
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 10 mg
Formel vikt 421.1638