Läs mer
Desmethyl Ketoprofen Methyl Ester, TRC
Produktkod. 30361120
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30361120 50 mg
1 options
Produktkod. 30361120 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29185050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desmethyl Ketoprofen Methyl Ester
CAS 24021-44-1
Molekylformel C16 H14 O3
Kvantitet 50 mg
Synonym 3-Benzoylbenzeneacetic Acid Methyl Ester, Acetic acid, (m-benzoylphenyl)-, methyl ester (8CI), Methyl (3-benzoylphenyl)acetate
InChI formel InChI=1S/C16H14O3/c1-19-15(17)11-12-6-5-9-14(10-12)16(18)13-7-3-2-4-8-13/h2-10H,11H2,1H3
LEDER COC(=O)Cc1cccc(c1)C(=O)c2ccccc2
IUPAC-namn methyl 2-(3-benzoylphenyl)acetate
Molekylvikt (g/mol) 254.28
Formel vikt 254.0943