Läs mer
Desmethyl Lacosamide, TRC
Produktkod. 30137359
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137359 25 mg
30011716 10 mg
30009173 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137359 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29183525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 175481-38-6
Molekylformel C12 H16 N2 O3
Molekylvikt (g/mol) 236.27
Synonym Propanamide, 2-(acetylamino)-3-hydroxy-N-(phenylmethyl)-, (R)-, (2R)-2-(Acetylamino)-3-hydroxy-N-(phenylmethyl)propanamide, (R)-N-Benzyl-2-acetamido-3-hydroxypropionamide, (2R)-2-(Acetylamino)-N-benzyl-3-hydroxypropanamide, Desmethyl Lacosamide, Lacosamide Imp. F (Pharmeuropa)
IUPAC-namn (2R)-2-acetamido-N-benzyl-3-hydroxypropanamide
LEDER CC(=O)N[C@H](CO)C(=O)NCc1ccccc1
Specifikationer
Kemiskt namn eller material Desmethyl Lacosamide
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C12H16N2O3/c1-9(16)14-11(8-15)12(17)13-7-10-5-3-2-4-6-10/h2-6,11,15H,7-8H2,1H3,(H,13,17)(H,14,16)/t11-/m1/s1
Formel vikt 236.1161
Procent renhet >95%