Läs mer
Desmethyl Metolazone, TRC
Produktkod. 30351835
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30351835 10 mg
1 options
Produktkod. 30351835 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29245010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desmethyl Metolazone
CAS 28524-40-5
Molekylformel C15H14ClN3O3S
Kvantitet 10 mg
Synonym 7-Chloro-1,2,3,4-tetrahydro-2-methyl-4-oxo-3-phenyl-6-quinazolinesulfonamide
InChI formel InChI=1S/C15H14ClN3O3S/c1-9-18-13-8-12(16)14(23(17,21)22)7-11(13)15(20)19(9)10-5-3-2-4-6-10/h2-9,18H,1H3,(H2,17,21,22)
LEDER CC1Nc2cc(Cl)c(cc2C(=O)N1c3ccccc3)S(=O)(=O)N
Molekylvikt (g/mol) 351.81