Läs mer
Desmethyl-O-Benzyl Apixaban Carboxylate, TRC
Produktkod. 30374039
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30374039 50 mg
1 options
Produktkod. 30374039 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE70287050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desmethyl-O-Benzyl Apixaban Carboxylate
CAS 1620494-29-2
Molekylformel C33 H32 N4 O5
Kvantitet 50 mg
Synonym Apixaban Impurity D, Ethyl 1-(4-(Benzyloxy)phenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxylate
InChI formel InChI=1S/C33H32N4O5/c1-2-41-33(40)30-28-19-21-36(25-13-11-24(12-14-25)35-20-7-6-10-29(35)38)32(39)31(28)37(34-30)26-15-17-27(18-16-26)42-22-23-8-4-3-5-9-23/h3-5,8-9,11-18H,2,6-7,10,19-22H2,1H3
LEDER CCOC(=O)c1nn(c2ccc(OCc3ccccc3)cc2)c4C(=O)N(CCc14)c5ccc(cc5)N6CCCCC6=O
IUPAC-namn ethyl 7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-1-(4-phenylmethoxyphenyl)-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxylate
Molekylvikt (g/mol) 564.631
Formel vikt 564.237