Läs mer
Desmethyl-O-Benzyl Apixaban, TRC
Produktkod. 30365830
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30365830 10 mg
30359102 100 mg
30364719 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30365830 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD11984510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1620494-31-6
Molekylformel C31 H29 N5 O4
Molekylvikt (g/mol) 535.593
Synonym 1-(4-(Benzyloxy)phenyl)-7-oxo-6-(4-(2-oxopiperidin-1-yl)phenyl)-4,5,6,7-tetrahydro-1H-pyrazolo[3,4-c]pyridine-3-carboxamide
IUPAC-namn 7-oxo-6-[4-(2-oxopiperidin-1-yl)phenyl]-1-(4-phenylmethoxyphenyl)-4,5-dihydropyrazolo[3,4-c]pyridine-3-carboxamide
LEDER NC(=O)c1nn(c2ccc(OCc3ccccc3)cc2)c4C(=O)N(CCc14)c5ccc(cc5)N6CCCCC6=O
Specifikationer
Kemiskt namn eller material Desmethyl-O-Benzyl Apixaban
Kvantitet 10 mg
InChI formel InChI=1S/C31H29N5O4/c32-30(38)28-26-17-19-35(23-11-9-22(10-12-23)34-18-5-4-8-27(34)37)31(39)29(26)36(33-28)24-13-15-25(16-14-24)40-20-21-6-2-1-3-7-21/h1-3,6-7,9-16H,4-5,8,17-20H2,(H2,32,38)
Formel vikt 535.222