missing translation for 'onlineSavingsMsg'
Läs mer

Desmethyl Piroxicam (Piroxicam Impurity B), TRC

Produktkod. 30355022
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355022 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30355022 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2921855MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CAS 65897-46-3
Molekylformel C14 H11 N3 O4 S
Kvantitet 5 mg
Synonym 2H-1,2-Benzothiazine-3-carboxamide, 4-hydroxy-N-2-pyridinyl-, 1,1-dioxide, 4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Piroxicam Imp. B (EP), Desmethyl Piroxicam, Desmethylpiroxicam
InChI formel InChI=1S/C14H11N3O4S/c18-13-9-5-1-2-6-10(9)22(20,21)17-12(13)14(19)16-11-7-3-4-8-15-11/h1-8,17-18H,(H,15,16,19)
LEDER OC1=C(NS(=O)(=O)c2ccccc12)C(=O)Nc3ccccn3
IUPAC-namn 4-hydroxy-1,1-dioxo-N-pyridin-2-yl-2H-1λ^{6},2-benzothiazine-3-carboxamide
Molekylvikt (g/mol) 317.32
Formel vikt 317.047
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.