Läs mer
Desmethyl Piroxicam (Piroxicam Impurity B), TRC
Produktkod. 30363784
Change view
Kvantitet:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30363784 50 mg
30356047 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30363784 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29218550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
CAS 65897-46-3
Molekylformel C14 H11 N3 O4 S
Kvantitet 50 mg
Synonym 2H-1,2-Benzothiazine-3-carboxamide, 4-hydroxy-N-2-pyridinyl-, 1,1-dioxide, 4-Hydroxy-N-(pyridin-2-yl)-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Piroxicam Imp. B (EP), Desmethyl Piroxicam, Desmethylpiroxicam
InChI formel InChI=1S/C14H11N3O4S/c18-13-9-5-1-2-6-10(9)22(20,21)17-12(13)14(19)16-11-7-3-4-8-15-11/h1-8,17-18H,(H,15,16,19)
LEDER OC1=C(NS(=O)(=O)c2ccccc12)C(=O)Nc3ccccn3
IUPAC-namn 4-hydroxy-1,1-dioxo-N-pyridin-2-yl-2H-1λ^{6},2-benzothiazine-3-carboxamide
Molekylvikt (g/mol) 317.32
Formel vikt 317.047