Läs mer
Desmethyl Ranitidine, TRC
Produktkod. 30363715
Change view
Kvantitet:
1 mg
Produktkod. Kvantitet
30363715 1 mg
1 options
Produktkod. 30363715 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2921601MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Kemiskt namn eller material Desmethyl Ranitidine
CAS 66357-25-3
Molekylformel C12 H20 N4 O3 S
Kvantitet 1 mg
Synonym N-Methyl-N'-[2-[[[5-[(methylamino)methyl]-2-furanyl]methyl]thio]ethyl]-2-nitro-1,1-ethenediamine, Desmethylranitidine
InChI formel InChI=1S/C12H20N4O3S/c1-13-7-10-3-4-11(19-10)9-20-6-5-15-12(14-2)8-16(17)18/h3-4,8,13-15H,5-7,9H2,1-2H3
LEDER CNCc1oc(CSCCNC(=C[N+](=O)[O-])NC)cc1
IUPAC-namn 1-N-methyl-1-N'-[2-[[5-(methylaminomethyl)furan-2-yl]methylsulfanyl]ethyl]-2-nitroethene-1,1-diamine
Molekylvikt (g/mol) 300.38
Formel vikt 300.1256