Läs mer
Desmethylene Oxobexarotene-13C4 Methyl Ester, TRC
Produktkod. 30357556
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30357556 10 mg
1 options
Produktkod. 30357556 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29113710MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desmethylene Oxobexarotene-13C4 Methyl Ester
CAS 1391054-72-0
Molekylformel C2013C4H28O3
Kvantitet 10 mg
Synonym Methyl 4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]benzoate -13C4, 4-[(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)carbonyl]-benzoic Acid Methyl Ester -13C4
InChI formel InChI=1S/C24H28O3/c1-15-13-19-20(24(4,5)12-11-23(19,2)3)14-18(15)21(25)16-7-9-17(10-8-16)22(26)27-6/h7-10,13-14H,11-12H2,1-6H3/i2+1,3+1,4+1,5+1
LEDER [13CH3]C1([13CH3])CCC([13CH3])([13CH3])c2cc(C(=O)c3ccc(cc3)C(=O)OC)c(C)cc12
IUPAC-namn methyl 4-[3-methyl-5,5,8,8-tetra((1^{13}C)methyl)-6,7-dihydronaphthalene-2-carbonyl]benzoate
Molekylvikt (g/mol) 368.45
Formel vikt 368.2173