Läs mer
Desmethylnarcotine, TRC
Produktkod. 30359211
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359211 10 mg
30352496 5 mg
30348858 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359211 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29236510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 521-40-4
Molekylformel C21 H21 N O7
Molekylvikt (g/mol) 399.39
Synonym (S)-3-((R)-4-Hydroxy-6-methyl-5,6,7,8-tetrahydro-[1,3]dioxolo[4,5-g]isoquinolin-5-yl)-6,7-dimethoxyisobenzofuran-1(3H)-one
IUPAC-namn (3S)-3-[(5R)-4-hydroxy-6-methyl-7,8-dihydro-5H-[1,3]dioxolo[4,5-g]isoquinolin-5-yl]-6,7-dimethoxy-3H-isobenzofuran-1-one
LEDER COc1ccc2[C@H](OC(=O)c2c1OC)[C@@H]3N(C)CCc4cc5OCOc5c(O)c34
Specifikationer
Kemiskt namn eller material Desmethylnarcotine
Kvantitet 10 mg
InChI formel InChI=1S/C21H21NO7/c1-22-7-6-10-8-13-20(28-9-27-13)17(23)14(10)16(22)18-11-4-5-12(25-2)19(26-3)15(11)21(24)29-18/h4-5,8,16,18,23H,6-7,9H2,1-3H3/t16-,18+/m1/s1
Formel vikt 399.1318