missing translation for 'onlineSavingsMsg'
Learn More

Desmocollin 2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 13876-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16822284

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a member of the desmocollin protein subfamily. Desmocollins, along with desmogleins, are cadherin-like transmembrane glycoproteins that are major components of the desmosome. Desmosomes are cell-cell junctions that help resist shearing forces and are found in high concentrations in cells subject to mechanical stress. This gene is found in a cluster with other desmocollin family members on chromosome 18. Mutations in this gene are associated with arrhythmogenic right ventricular dysplasia-11, and reduced protein expression has been described in several types of cancer. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

Desmocollin 2
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P55292, Q02487
ARVD11, Cadherin family member 2, CDHF2, desmocollin 2, Desmocollin 3, Desmosomal glycoprotein II, Desmosomal glycoprotein III, DG2, DGII/III, DSC2, DSC3
Rabbit
IgG
150 μL
-20°C
Primary
13506, 1824
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
DSC2
Liquid
DSC2
Desmocollin 2 Fusion Protein Ag4956
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documents
Certificates
Special Offers

Special Offers