Läs mer
Desnitro-imidacloprid Hydrochloride, TRC
Produktkod. 30134347
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134347 25 mg
30138315 100 mg
17954285 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134347 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN56121525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 127202-53-3
Molekylformel C9 H11 Cl N4 . Cl H
Molekylvikt (g/mol) 247.12
Synonym 1H-Imidazol-2-amine, 1-[(6-chloro-3-pyridinyl)methyl]-4,5-dihydro-, hydrochloride (1:1), 1H-Imidazol-2-amine, 1-[(6-chloro-3-pyridinyl)methyl]-4,5-dihydro-, monohydrochloride (9CI), 1-[(6-Chloro-3-pyridinyl)methyl]-4,5-dihydro-1H-imidazol-2-amine hydrochloride (1:1), Imidacloprid-guanidine hydrochloride, Desnitro-imidacloprid hydrochloride
IUPAC-namn 1-[(6-chloropyridin-3-yl)methyl]-4,5-dihydroimidazol-2-amine;hydrochloride
LEDER Cl.NC1=NCCN1Cc2ccc(Cl)nc2
Specifikationer
Kemiskt namn eller material Imidacloprid-guanidine hydrochloride
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C9H11ClN4.ClH/c10-8-2-1-7(5-13-8)6-14-4-3-12-9(14)11;/h1-2,5H,3-4,6H2,(H2,11,12);1H
Formel vikt 246.0439
Procent renhet >95%