Läs mer
Desnitro-olefin Imidacloprid, TRC
Produktkod. 30480357
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30480357 25 mg
1 options
Produktkod. 30480357 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN56122025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Imidacloprid-desnitro-olefin
CAS 187022-17-9
Molekylformel C9 H9 Cl N4
Kvantitet 25 mg
Synonym Imidacloprid-desnitro-olefin, 1-[(6-Chloro-3-pyridinyl)methyl]-1H-imidazol-2-amine (ACI), 1H-Imidazol-2-amine','1-[(6-chloro-3-pyridinyl)methyl]- (9CI','ACI)
InChI formel InChI=1S/C9H9ClN4/c10-8-2-1-7(5-13-8)6-14-4-3-12-9(14)11/h1-5H,6H2,(H2,11,12)
LEDER Nc1nccn1Cc2ccc(Cl)nc2
IUPAC-namn 1-[(6-chloropyridin-3-yl)methyl]imidazol-2-amine
Molekylvikt (g/mol) 208.648
Formel vikt 208.052