Läs mer
Desthiazolylmethyloxycarbonyl Ritonavir, TRC
Produktkod. 30358742
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30358742 5 mg
1 options
Produktkod. 30358742 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2974305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desthiazolylmethyloxycarbonyl ritonavir
CAS 176655-55-3
Molekylformel C32 H45 N5 O3 S
Kvantitet 5 mg
Synonym Butanamide, N-[(1S,3S,4S)-4-amino-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-3-methyl-2-[[[methyl[[2-(1-methylethyl)-4-thiazolyl]methyl]amino]carbonyl]amino]-, (2S)-, Butanamide, N-[4-amino-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-3-methyl-2-[[[methyl[[2-(1-methylethyl)-4-thiazolyl]methyl]amino]carbonyl]amino]-, [1S-[1R*(R*),3R*,4R*]]-, (2S)-N-[(1S,3S,4S)-4-Amino-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-3-methyl-2-[[[methyl[[2-(1-methylethyl)-4-thiazolyl]methyl]amino]carbonyl]amino]butanamide
InChI formel InChI=1S/C32H45N5O3S/c1-21(2)29(36-32(40)37(5)19-26-20-41-31(35-26)22(3)4)30(39)34-25(16-23-12-8-6-9-13-23)18-28(38)27(33)17-24-14-10-7-11-15-24/h6-15,20-22,25,27-29,38H,16-19,33H2,1-5H3,(H,34,39)(H,36,40)/t25-,27-,28-,29-/m0/s1
LEDER CC(C)[C@H](NC(=O)N(C)Cc1csc(n1)C(C)C)C(=O)N[C@H](C[C@H](O)[C@@H](N)Cc2ccccc2)Cc3ccccc3
IUPAC-namn (2S)-N-[(2S,4S,5S)-5-amino-4-hydroxy-1,6-diphenylhexan-2-yl]-3-methyl-2-[[methyl-[(2-propan-2-yl-1,3-thiazol-4-yl)methyl]carbamoyl]amino]butanamide
Molekylvikt (g/mol) 579.8
Formel vikt 579.3243