Läs mer
Destriazolyl Bromo Propiconazole (Mixture of Diastereomers), TRC
Produktkod. 17983045
Change view
Kvantitet:
250 mg
2.5 g
Förpackningsstorlek:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17983045 2.5 g 2.5g
17931765 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17983045 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2978252.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60207-89-8
Molekylformel C13 H15 Br Cl2 O2
Molekylvikt (g/mol) 354.07
Synonym 2-(Bromomethyl)-2-(2,4-dichlorophenyl)-4-propyl-1,3-dioxolane, 2-Bromomethyl-2-(2,4-dichlorophenyl)-4-propyldioxolane;
IUPAC-namn 2-(bromomethyl)-2-(2,4-dichlorophenyl)-4-propyl-1,3-dioxolane
LEDER CCCC1COC(CBr)(O1)c2ccc(Cl)cc2Cl
Specifikationer
Kemiskt namn eller material Destriazolyl Bromo Propiconazole (Mixture of Diastereomers)
Kvantitet 2.5 g
InChI formel InChI=1S/C13H15BrCl2O2/c1-2-3-10-7-17-13(8-14,18-10)11-5-4-9(15)6-12(11)16/h4-6,10H,2-3,7-8H2,1H3
Formel vikt 351.9632