Läs mer
Desulfo Oxo-ambazone, TRC
Produktkod. 30357114
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30357114 100 mg
1 options
Produktkod. 30357114 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD297875100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Desulfo Oxo-ambazone
CAS 61566-10-7
Molekylformel C8H11N7O
Kvantitet 100 mg
Synonym (4-Oxo-2,5-cyclohexadien-1-ylideneamino)guanidine Semicarbazone, 2-[4-[(Aminoiminomethyl)hydrazono]-2,5-cyclohexadien-1-ylidene]-hydrazinecarboxamide, 2-[4-[2-(Aminoiminomethyl)hydrazinylidene]-2,5-cyclohexadien-1-ylidene]hydrazinecarboxamide
InChI formel InChI=1S/C8H11N7O/c9-7(10)14-12-5-1-3-6(4-2-5)13-15-8(11)16/h1-4,13H,(H3,9,10)(H3,11,15,16)
LEDER NC(=N)NN=C1C=CC(=NNC(=O)N)C=C1
Molekylvikt (g/mol) 221.22
Formel vikt 221.1025