missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Devarda's Alloy, granular

Catalog Number p-4408793
Click to view available options
Quantity:
100 g
500 g
8049-11-4
Al9Cu10Zn
943.67
MFCD00198176
MPKKOQSNGWAMJD-UHFFFAOYSA-N
devarda's alloy, devarda's alloy, granular, al.cu.zn, devarda's alloy, p.a., alumane copper zinc, devarda's alloy, puriss. p.a., devarda's alloy, saj special grade, devarda's alloy, powder,-100 mesh, devarda's alloy, filings, +100 mesh, devarda inverted exclamation mark s alloy
16211762
aluminum;copper;zinc
[Al].[Cu].[Zn]
8049-11-4
Al9Cu10Zn
943.67
MFCD00198176
MPKKOQSNGWAMJD-UHFFFAOYSA-N
devarda's alloy, devarda's alloy, granular, al.cu.zn, devarda's alloy, p.a., alumane copper zinc, devarda's alloy, puriss. p.a., devarda's alloy, saj special grade, devarda's alloy, powder,-100 mesh, devarda's alloy, filings, +100 mesh, devarda inverted exclamation mark s alloy
16211762
aluminum;copper;zinc
[Al].[Cu].[Zn]

Devarda's Alloy is used as reducing agent in analytical chemistry for the determination of nitrates after their reduction to ammonia under alkaline conditions. It is also used in the quantitative or qualitative analysis of nitrates in agriculture and soil science before the development of ion chromatography. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 4.355
Quantity 100 g
Solubility Information Insoluble in water. Slightly soluble in hydrochloric acid
Form Granular
Chemical Name or Material Devarda's Alloy

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'