Läs mer
Dexamethasone 17-Propionate, TRC
Produktkod. 30358926
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30358926 1 mg
30350711 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30358926 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2988201MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15423-89-9
Molekylformel C25 H33 F O6
Molekylvikt (g/mol) 448.52
Synonym (11β,16α)-9-Fluoro-11,21-dihydroxy-16-methyl-17-(1-oxopropoxy)-pregna-1,4-diene-3,20-dione, 9-Fluoro-11β,17,21-trihydroxy-16α-methyl-pregna-1,4-diene-3,20-dione 17-Propionate
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
LEDER CCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
Specifikationer
Kemiskt namn eller material Dexamethasone 17-propionate
Kvantitet 1 mg
InChI formel InChI=1S/C25H33FO6/c1-5-21(31)32-25(20(30)13-27)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,26)19(29)12-23(18,25)4/h8-9,11,14,17-19,27,29H,5-7,10,12-13H2,1-4H3/t14-,17+,18+,19+,22+,23+,24+,25+/m1/s1
Formel vikt 448.2261