Läs mer
Dexamethasone 21-Palmitate, TRC
Produktkod. 17973045
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17973045 50 mg
17931575 250 mg
17963165 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17973045 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29883050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 14899-36-6
Molekylformel C38 H59 F O6
Molekylvikt (g/mol) 630.87
Synonym (11β,16α)-9-Fluoro-11,17-dihydroxy-16-methyl-21-[(1-oxohexadecyl)oxy]pregna-1,4-diene-3,20-dione, Dexamethasone Palmitate, Limethason, Limethasone, Lipotalon;
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] hexadecanoate
LEDER CCCCCCCCCCCCCCCC(=O)OCC(=O)[C@@]1(O)[C@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Dexamethasone 21-Palmitate
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C38H59FO6/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-34(43)45-26-33(42)38(44)27(2)23-31-30-20-19-28-24-29(40)21-22-35(28,3)37(30,39)32(41)25-36(31,38)4/h21-22,24,27,30-32,41,44H,5-20,23,25-26H2,1-4H3/t27-,30+,31+,32+,35+,36+,37+,38+/m1/s1
Formel vikt 630.4296
Procent renhet >95%