Läs mer
Dexamethasone Acetate Impurity E, TRC
Produktkod. 17982335
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982335 25 mg
17934905 5 mg
17942865 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17982335 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29875525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1524-94-3
Molekylformel C24 H33 F O6
Molekylvikt (g/mol) 436.51
Synonym 9-Fluoro-11β,17-dihydroxy-16α-methyl-3,20-dioxopregn-4-en-21-yl Acetate, Dexamethasone Acetate Imp. E (EP), Pregn-4-ene-3,20-dione, 21-(acetyloxy)-9-fluoro-11,17-dihydroxy-16-methyl-, (11β,16α)- (9CI), Corticosterone, 9-fluoro-17-hydroxy-16α-methyl-, 21-acetate (6CI), Pregn-4-ene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-acetate (7CI,8CI), 16α-Methyl-9α-fluorocortisol 21-acetate, 9-Fluoro-11β,17,21-trihydroxy-16α-methylpregn-4-ene-3,20-dione, 21-acetate
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C
Specifikationer
Kemiskt namn eller material 9-Fluoro-11beta,17-dihydroxy-16alpha-methyl-3,20-dioxopregn-4-en-21-yl Acetate
Kvantitet 25 mg
InChI formel InChI=1S/C24H33FO6/c1-13-9-18-17-6-5-15-10-16(27)7-8-21(15,3)23(17,25)19(28)11-22(18,4)24(13,30)20(29)12-31-14(2)26/h10,13,17-19,28,30H,5-9,11-12H2,1-4H3/t13-,17+,18+,19+,21+,22+,23+,24+/m1/s1
Formel vikt 436.2261