Läs mer
Dexamethasone-biotin, TRC
Produktkod. 30359514
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30359514 10 mg -
30361753 2.5 mg 2.5mg
30361837 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359514 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29876510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 85679-56-7
Molekylformel C36 H53 F N4 O6 S
Molekylvikt (g/mol) 688.89
Synonym N-[5-[[[(11β,16α,17α)-9-Fluoro-11,17-dihydroxy-16-methyl-3-oxoandrosta-1,4-dien-17-yl]carbonyl]amino]pentyl]hexahydro-2-oxo-1H-thieno[3,4-d]imidazole-4-pentanamide
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R,17R)-N-[5-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]pentyl]-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxamide
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)NCCCCCNC(=O)CCCC[C@@H]5SC[C@@H]6NC(=O)N[C@H]56
Specifikationer
Kemiskt namn eller material Dexamethasone-biotin
Kvantitet 10 mg
InChI formel InChI=1S/C36H53FN4O6S/c1-21-17-25-24-12-11-22-18-23(42)13-14-33(22,2)35(24,37)28(43)19-34(25,3)36(21,47)31(45)39-16-8-4-7-15-38-29(44)10-6-5-9-27-30-26(20-48-27)40-32(46)41-30/h13-14,18,21,24-28,30,43,47H,4-12,15-17,19-20H2,1-3H3,(H,38,44)(H,39,45)(H2,40,41,46)/t21-,24+,25+,26+,27+,28+,30+,33+,34+,35+,36+/m1/s1
Formel vikt 688.367