Läs mer
Dexamethasone-Delta17,20 21-Aldehyde, TRC
Produktkod. 30040938
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30040938 50 mg
30040394 10 mg
30041148 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30040938 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29879050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6762-51-2
Molekylformel C22 H27 F O4
Molekylvikt (g/mol) 374.45
Synonym (11β,16α)-9-Fluoro-11,20-dihydroxy-16-methyl-3-oxopregna-1,4,17(20)-trien-21-al, 9-Fluoro-11β,20-dihydroxy-16α-methyl-3-oxopregna-1,4,17(20)-trien-21-al
IUPAC-namn 2-(9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-3,6,7,8,9,10,11,12,13,14,15,16-dodecahydro-17H-cyclopenta[a]phenanthren-17-ylidene)-2-hydroxyacetaldehyde
LEDER F[C@@]12[C@]([C@]3([C@](C)(C[C@@H]1O)C(=C(C=O)O)[C@H](C)C3)[H])(CCC=4[C@]2(C)C=CC(=O)C4)[H]
Specifikationer
Kemiskt namn eller material (E/Z)-Dexamethasone-delta17,20 21-aldehyde
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C22H27FO4/c1-12-8-16-15-5-4-13-9-14(25)6-7-21(13,3)22(15,23)18(27)10-20(16,2)19(12)17(26)11-24/h6-7,9,11-12,15-16,18,26-27H,4-5,8,10H2,1-3H3/t12-,15+,16+,18+,20+,21+,22+/m1/s1
Formel vikt 374.1893
Procent renhet >95%