Läs mer
Dexamethasone Dipropionate, TRC
Produktkod. 30005545
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005545 5 mg
30004277 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005545 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2988155MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55541-30-5
Molekylformel C28 H37 F O7
Molekylvikt (g/mol) 504.59
Synonym (11β,16α)-9-Fluoro-11-hydroxy-16-methyl-17,21-bis(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Dexamethasone 17,21-dipropionate, Dexamethasone dipropionate, Methaderm, ST 12, THS 101
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
LEDER CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Dexamethasone Dipropionate
Kvantitet 5 mg
InChI formel InChI=1S/C28H37FO7/c1-6-23(33)35-15-22(32)28(36-24(34)7-2)16(3)12-20-19-9-8-17-13-18(30)10-11-25(17,4)27(19,29)21(31)14-26(20,28)5/h10-11,13,16,19-21,31H,6-9,12,14-15H2,1-5H3/t16-,19+,20+,21+,25+,26+,27+,28+/m1/s1
Formel vikt 504.2523