missing translation for 'onlineSavingsMsg'
Läs mer

Dexamethasone Hemisuccinate Antibody, Novus Biologicals™

Produktkod. 18058953 Handla allt Bio Techne Produkter
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
0.1 mL
0.5 mL
Förpackningsstorlek:
0.1ml
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18058953 0.1 mL 0.1ml
18020694 0.5 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 18058953 Leverantör Novus Biologicals Leverantörsnummer NB100625070.1ML

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Dexamethasone Hemisuccinate Polyclonal specifically detects Dexamethasone Hemisuccinate in Non-species specific samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Dexamethasone Hemisuccinate
Användningsområden ELISA
Klassificering Polyclonal
Konjugera Unconjugated
Utspädning ELISA 1:100-1:2000
Formulering PBS with 0.09% Sodium Azide
Värd art Rabbit
Immunogen Dexamethasone conjugated to Bovine Serum Albumin.
Reningsmetod Affinity Purified
Kvantitet 0.1 mL
Regulatorisk status RUO
Primär eller sekundär Primary
Testspecificitet This antibody recognises Dexamethasone hemisuccinate, a synthetic steroid hormone, often used as an immunosupressant or anti-inflammatory. This antibody exhibits the following cross-reactivities: Dexamethasone 100% 5 Dexamethasone-21-Ac 29% Beta-Methasone 0.1% Dexamethasone-21-Phosphate <0.01% Testosterone <0.01%
Målarter Non-species specific
Innehåll och lagring Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotyp IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.