Läs mer
Dexamethasone Hemisuccinate, TRC
Produktkod. 30356069
Change view
Kvantitet:
50 mg
100 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356069 100 mg
30356303 1 g
30348791 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30356069 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD298855100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3800-86-0
Molekylformel C26 H33 F O8
Molekylvikt (g/mol) 492.53
Synonym (11β,16α)-21-(3-Carboxy-1-oxopropoxy)-9-fluoro-11,17-dihydroxy-16-methyl-pregna-1,4-diene-3,20-dione
IUPAC-namn 4-[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxo-ethoxy]-4-oxo-butanoic acid
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)CCC(=O)O
Specifikationer
Kemiskt namn eller material Dexamethasone Hemisuccinate
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C26H33FO8/c1-14-10-18-17-5-4-15-11-16(28)8-9-23(15,2)25(17,27)19(29)12-24(18,3)26(14,34)20(30)13-35-22(33)7-6-21(31)32/h8-9,11,14,17-19,29,34H,4-7,10,12-13H2,1-3H3,(H,31,32)/t14-,17+,18+,19+,23+,24+,25+,26+/m1/s1
Formel vikt 492.2159
Procent renhet >95%