missing translation for 'onlineSavingsMsg'
Läs mer

Dexamethasone Oxetanone, TRC

Produktkod. 30355438
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355438 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Product Code. 30355438 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCD29887510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Dexamethasone oxetanone
CAS 4089-36-5
Molekylformel C22 H27 F O4
Kvantitet 10 mg
Synonym Pregna-1,4-diene-3,20-dione, 17,21-epoxy-9-fluoro-11-hydroxy-16-methyl-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 17,21-epoxy-9-fluoro-11β-hydroxy-16α-methyl- (7CI), (11β,16α)-17,21-Epoxy-9-fluoro-11-hydroxy-16-methylpregna-1,4-diene-3,20-dione, Dexamethasone oxetanone
InChI formel InChI=1S/C22H27FO4/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12)18(26)11-27-22/h6-7,9,12,15-17,25H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@]15OCC5=O
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11-hydroxy-10,13,16-trimethylspiro[6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17,2'-oxetane]-3,3'-dione
Molekylvikt (g/mol) 374.45
Formel vikt 374.1893
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.