Läs mer
Dexamethasone Sodium Sulfobenzoate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Dexamethasone Sodium Sulfobenzoate |
| CAS | 3936-02-5 |
| Molekylformel | C29 H32 F O9 S . Na |
| Kvantitet | 50 mg |
| Synonym | 9-Fluoro-11,17-dihydroxy-16-methyl-21-[(3-sulfobenzoyl)oxy]pregna-1,4-diene-3,20-dione Sodium Salt (1:1), Pregna-1,4-diene-3,20-dione, 9-fluoro-11,17-dihydroxy-16-methyl-21-[(3-sulfobenzoyl)oxy]-, monosodium salt, (11β,16α)- (9CI), Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-(m-sulfobenzoate), monosodium salt (8CI), Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 21-m-sulfobenzoate, sodium salt (7CI), 21-(3-Sulfobenzoyloxy)dexamethasone sodium salt, Dexamethasone 21-sodium-m-sulfobenzoate, Dexamethasone m-sulfobenzoate sodium, Dexamethasone metasulfobenzoate sodium, Dexamethasone sodium m-sulfobenzoate, Dexamethasone sulfobenzoate sodium, Sisotek, Sodium dexamethasone-m-sulfobenzoate, Dexamethasone-21-sodium-m-Sulfobenzoate, Sisotek, 21-(3-Sulfobenzoyloxy)dexamethasone Sodium Salt, Sodium 3-((2-((8S,9R,10S,11S,13S,14S,16R,17R)-9-Fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,9,10,11,12,13,14,15,16,17-dodecahydro-3H-cyclopenta[a]phenanthren-17-yl)-2-oxoethoxy)carbonyl)benzenesulfonate, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-Pregna-1,4-diene-3,20-dione-21-m-sulfobenzoate Sodium Salt |
| InChI formel | InChI=1S/C29H33FO9S.Na/c1-16-11-22-21-8-7-18-13-19(31)9-10-26(18,2)28(21,30)23(32)14-27(22,3)29(16,35)24(33)15-39-25(34)17-5-4-6-20(12-17)40(36,37)38;/h4-6,9-10,12-13,16,21-23,32,35H,7-8,11,14-15H2,1-3H3,(H,36,37,38);/q;+1/p-1/t16-,21+,22+,23+,26+,27+,28+,29+;/m1./s1 |
| LEDER | [Na+].C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)COC(=O)c5cccc(c5)S(=O)(=O)[O-] |
| IUPAC-namn | sodium;3-[2-[(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethoxy]carbonylbenzenesulfonate |
| Molekylvikt (g/mol) | 598.6122 |
| Formel vikt | 598.1649 |