Läs mer
Dexamethasone Valerate, TRC
Produktkod. 18003650
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18003650 10 mg -
18060950 2.5 mg 2.5mg
18030250 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18003650 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29887010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 33755-46-3
Molekylformel C27 H37 F O6
Molekylvikt (g/mol) 476.58
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]-, (11β,16α)-, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16α-methyl-, 17-valerate (8CI), (11β,16α)-9-Fluoro-11,21-dihydroxy-16-methyl-17-[(1-oxopentyl)oxy]pregna-1,4-diene-3,20-dione, DV 17, Dexamethasone 17-valerate, Dexamethasone valerate, Betamethasone Valerate Imp. C (EP)
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] pentanoate
LEDER CCCCC(=O)O[C@@]1([C@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
Specifikationer
Kemiskt namn eller material Dexamethasone Valerate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19+,20+,21+,24+,25+,26+,27+/m1/s1
Formel vikt 476.2574
Procent renhet >95%