Läs mer
Dexmedetomidine Hydrochloride, TRC
Produktkod. 30134867
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134867 100 mg
30134137 50 mg
30011663 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134867 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD299008100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 145108-58-3
Molekylformel C13 H16 N2 . Cl H
Molekylvikt (g/mol) 236.74
Synonym Dexmedetomidine hydrochloride, 1H-Imidazole, 5-[(1S)-1-(2,3-dimethylphenyl)ethyl]-, hydrochloride (1:1), 1H-Imidazole, 4-[(1S)-1-(2,3-dimethylphenyl)ethyl]-, monohydrochloride (9CI), 1H-Imidazole, 4-[1-(2,3-dimethylphenyl)ethyl]-, monohydrochloride, (S)-, 5-[(1S)-1-(2,3-Dimethylphenyl)ethyl]-1H-imidazole hydrochloride, (S)-4-(1-(2,3-Dimethylphenyl)ethyl)-1H-imidazole hydrochloride, Dex, Dexdomitor, Dexmetomidine, Precedex
IUPAC-namn 4-[(1S)-1-(2,3-dimethylphenyl)ethyl]-1H-imidazole;hydrochloride
LEDER Cl.C[C@H](c1c[nH]cn1)c2cccc(C)c2C
Specifikationer
Kemiskt namn eller material Dexmedetomidine Hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C13H16N2.ClH/c1-9-5-4-6-12(10(9)2)11(3)13-7-14-8-15-13;/h4-8,11H,1-3H3,(H,14,15);1H/t11-;/m0./s1
Formel vikt 236.108