Läs mer
Dexon, TRC
Produktkod. 30355215
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30355215 100 mg
30360012 50 mg
30348963 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30355215 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD299603100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 140-56-7
Molekylformel C8 H10 N3 O3 S . Na
Molekylvikt (g/mol) 251.24
Synonym Diazenesulfonic acid, 2-[4-(dimethylamino)phenyl]-, sodium salt (1:1) (ACI), Benzenediazosulfonic acid, p-(dimethylamino)-, sodium salt (6CI, 7CI, 8CI), Diazenesulfonic acid, [4-(dimethylamino)phenyl]-, sodium salt (9CI), 4-Dimethylaminobenzendiasulfonic acid sodium salt, BAY 22555, BAY 5072, Bayer 22555, Bayer 5072, DAPA, DAPA (pesticide), Deksonal, Dexon, Dexon (pesticide), Dexon 70, Dexoxon, Fenaminosulf, Lesan, p-(Dimethylamino)benzenediazosulfonic acid sodium salt, p-Diazodimethylaniline sodium sulfonate, sodium (E)-[4-(dimethylamino)phenyl]diazenesulfonate, Sodium 4-(dimethylamino)benzenediazosulfonate, sodium 4-dimethylaminobenzenediazosulphonate, Sodium [4-(dimethylamino)phenyl]diazenesulfonate, Sodium p-(dimethylamino)benzenediazosulfonate
IUPAC-namn sodium;N-[4-(dimethylamino)phenyl]iminosulfamate
LEDER [Na+].CN(C)c1ccc(cc1)N=NS(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Fenaminosulf
Kvantitet 100 mg
InChI formel InChI=1S/C8H11N3O3S.Na/c1-11(2)8-5-3-7(4-6-8)9-10-15(12,13)14;/h3-6H,1-2H3,(H,12,13,14);/q;+1/p-1
Formel vikt 251.0341