Läs mer
Dexrazoxane, TRC
Produktkod. 30359795
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30359795 250 mg
30356168 50 mg
30360981 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30359795 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD299050250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24584-09-6
Molekylformel C11 H16 N4 O4
Molekylvikt (g/mol) 268.27
Synonym 2,6-Piperazinedione, 4,4'-[(1S)-1-methyl-1,2-ethanediyl]bis-, 2,6-Piperazinedione, 4,4'-(1-methyl-1,2-ethanediyl)bis-, (S)-, 2,6-Piperazinedione, 4,4'-propylenedi-, (+)- (8CI), 4,4'-[(1S)-1-Methyl-1,2-ethanediyl]bis[2,6-piperazinedione], (+)-1,2-Bis(3,5-dioxopiperazin-1-yl)propane, (S)-(+)-1,2-Bis(3,5-dioxopiperazin-1-yl)propane, ADR 529, Dexrazoxane, ICRF 187, NSC 169780, Savene, Totect, Zinecard
IUPAC-namn 4-[(2S)-2-(3,5-dioxopiperazin-1-yl)propyl]piperazine-2,6-dione
LEDER C[C@@H](CN1CC(=O)NC(=O)C1)N2CC(=O)NC(=O)C2
Specifikationer
Kemiskt namn eller material Dexrazoxane
Kvantitet 250 mg
InChI formel InChI=1S/C11H16N4O4/c1-7(15-5-10(18)13-11(19)6-15)2-14-3-8(16)12-9(17)4-14/h7H,2-6H2,1H3,(H,12,16,17)(H,13,18,19)/t7-/m0/s1
Formel vikt 268.1172