Learn More
Dextran sulfate sodium salt, MW ca 8,000, Thermo Scientific Chemicals
Description
Dextran sulfate is used asa nucleic acid hybridization accelerant. The ability of the polymer to precipitate fibrinogen and low-density lipoproteins from plasma solutions has been used for analysis and for anticoagulant applications. Dextran Sulfate Sodium (DSS) which has proven to have versatile value in Biomedical/clinical research. DSS can be used to study Colitis (Inflammatory Bowel disease) as well as an antiviral as agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White to Yellow |
Physical Form | Powder |
Merck Index | 14,2951 |
Solubility Information | Soluble in water (100mg/ml). |
Quantity | 25 g |
Formula Weight | ≈8 kd |
Odor | Phenol-like |
Chemical Name or Material | Dextran sulfate sodium salt |
RUO – Research Use Only