Läs mer
Dextromethorphan, Lipomed
Produktkod. 30460252
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30460252 10 mg
1 options
Produktkod. 30460252 Leverantör Lipomed Leverantörsnummer LPMDEX873FB10
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Dextromethorphan
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Dextromethorphan
CAS 125-71-3
Analyt- eller komponentnamn Dextromethorphan
Frakt skick Room Temperature
Molekylformel C18 H25 N O
Synonym Morphinan, 3-methoxy-17-methyl-, (9α,13α,14α)-, 9α,13α,14α-Morphinan, 3-methoxy-17-methyl- (8 CI), (9α,13α,14α)-3-Methoxy-17-methylmorphinan, (+)-3-Methoxy-17-methylmorphinan, (+)-Dextromethorphan, 3-Methoxy-17-methyl-9α,13α,14α-morphinan, Ba 2666, DEX, Dextromethorphan, Nodex, d-Methorphan
InChI formel InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
LEDER COc1ccc2C[C@H]3 [C@H]4 CCCC[C@@]4(CCN3C)c2c1
IUPAC-namn (1 S,9 S,10 S)-4-methoxy-17-methyl-17-azatetracyclo[7.5.3.01,10.02,7 ]heptadeca-2(7),3,5-triene
Industry Type Pharmaceutical
Molekylvikt (g/mol) 271.397
Kvantitet 10 mg
Formel vikt 271.194 g/mol