Läs mer
Dextrorphan Tartrate Salt, TRC
Produktkod. 17991105
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17991105 500 mg
17922085 100 mg
17972085 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17991105 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD299485500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 143-98-6
Molekylformel C17 H23 N O . C4 H6 O6
Molekylvikt (g/mol) 407.46
Synonym ent-17-Methylmorphinan-3-ol (L)-Tartrate, Dextromethorphan Hydrobromide Imp. B (EP) as (L)-Tartrate, Dextrorphan Tartrate, O-Desmethyldextromethorphan Tartrate, Morphinan-3-ol, 17-methyl-, (9α,13α,14α)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1), Morphinan-3-ol, 17-methyl-, (9α,13α,14α)-, (2R,3R)-2,3-dihydroxybutanedioate (1:1) (salt) (9CI), Morphinan-3-ol, 17-methyl-, (9α,13α,14α)-, [R-(R*,R*)]-2,3-dihydroxybutanedioate (1:1) (salt), Dextrorphan bitartrate, NIH 4591, d-3-Hydroxy-N-methylmorphinan tartrate
IUPAC-namn (9α,13α,14α)-3-Hydroxy-17-methylmorphinan (2R,3R)-2,3-dihydroxybutanoate
LEDER CN1CC[C@@]23CCCC[C@@H]2[C@@H]1Cc4ccc(O)cc34.O[C@H]([C@@H](O)C(=O)O)C(=O)O
Specifikationer
Kemiskt namn eller material Dextrorphan tartrate
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C17H23NO.C4H6O6/c1-18-9-8-17-7-3-2-4-14(17)16(18)10-12-5-6-13(19)11-15(12)17;5-1(3(7)8)2(6)4(9)10/h5-6,11,14,16,19H,2-4,7-10H2,1H3;1-2,5-6H,(H,7,8)(H,9,10)/t14-,16+,17+;1-,2-/m11/s1
Formel vikt 407.1944
Procent renhet >95%