missing translation for 'onlineSavingsMsg'
Learn More

Dextrose U.S.P. Granular, Macron Fine Chemicals™

Catalog Number 15568314
Click to view available options
POLYSTORMOR; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 50-99-7
Packaging POLYSTORMOR
Molecular Formula C6H12O6
Quantity 500 g
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
SMILES C(C(C(C(C(C=O)O)O)O)O)O
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
Molecular Weight (g/mol) 180.156
PubChem CID 107526
ChEBI CHEBI:42758
Formula Weight 180.16
Grade USP
Physical Form Granules
Chemical Name or Material Dextrose, Anhydrous
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'