Läs mer
Dextrosimendan 6-Phenyl, TRC
Produktkod. 30476667
Change view
Kvantitet:
25 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30476667 25 mg
30470325 250 mg
30474398 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30476667 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL58917025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2290570-69-1
Molekylformel C11H12N2O
Molekylvikt (g/mol) 188.23
Synonym (5S)-4,5-Dihydro-5-methyl-6-phenyl-3(2H)-pyridazinone
IUPAC-namn (4S)-4-methyl-3-phenyl-4,5-dihydro-1H-pyridazin-6-one
LEDER C[C@H]1CC(=O)NN=C1c2ccccc2
Specifikationer
Kemiskt namn eller material Dextrosimendan 6-Phenyl
Kvantitet 25 mg
InChI formel InChI=1S/C11H12N2O/c1-8-7-10(14)12-13-11(8)9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,12,14)/t8-/m0/s1
Formel vikt 188.095