missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGKA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | DGKA |
---|---|
Concentration | 0.17 mg/mL |
Applications | Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838703
|
Proteintech
11547-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828703
|
Proteintech
11547-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024
Log in to see stock. |
|||||
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.Specifications
DGKA | |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O88673, P23743, P51556 | |
13139, 140866, 1606 | |
DGKA Fusion Protein Ag2075 | |
Primary | |
-20°C | |
DGKA |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
80 kDa diacylglycerol kinase, DAG kinase alpha, DAGK, DAGK1, DGK alpha, DGKA, Diacylglycerol kinase alpha, Diglyceride kinase alpha | |
Dgka | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |