missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DGKA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11547-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the eukaryotic diacylglycerol kinase family. It acts as a modulator that competes with protein kinase C for the second messenger diacylglycerol in intracellular signaling pathways. It also plays an important role in the resynthesis of phosphatidylinositols and phosphorylating diacylglycerol to phosphatidic acid. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.Specifications
DGKA | |
Polyclonal | |
Unconjugated | |
DGKA | |
80 kDa diacylglycerol kinase, DAG kinase alpha, DAGK, DAGK1, DGK alpha, DGKA, Diacylglycerol kinase alpha, Diglyceride kinase alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
13139, 140866, 1606 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88673, P23743, P51556 | |
Dgka | |
DGKA Fusion Protein Ag2075 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |