Läs mer
(+)-DHMEQ, MedChemExpress
Produktkod. 16496118
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16496118 10 mg
16490216 2 mg
16488717 5 mg
16443436 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16496118 Leverantör Medchem Express Leverantörsnummer HY14645A
MedChemExpress (+)-DHMEQ is an activator of antioxidant transcription factor Nrf2. (+)-DHMEQ is the enantiomer of (-)-DHMEQ. (-)-DHMEQ inhibits NF-kB than its enantiomer (+)-DHMEQ.
(+)-DHMEQ is an activator of antioxidant transcription factor Nrf2. (+)-DHMEQ is the enantiomer of (-)-DHMEQ. (-)-DHMEQ inhibits NF-kB than its enantiomer (+)-DHMEQ.
Specifikationer
Kemiskt namn eller material (+)-DHMEQ
CAS 287194-41-6
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C13H11NO5
Kvantitet 10 mg
Synonym (1R,2R,6R)-Dehydroxymethylepoxyquinomicin (1R,2R,6R)-DHMEQ
Löslighetsinformation DMSO : 100 mg/mL (382.80 mM; Need ultrasonic)
LEDER O=C(NC([C@@H](O)[C@@]1([H])O[C@@]21[H])=CC2=O)C3=CC=CC=C3O
Molekylvikt (g/mol) 261.23
Formel vikt 261.23
Procent renhet 99.65%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation