Läs mer
DHPMA, TRC
Produktkod. 30134323
Change view
Kvantitet:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134323 500 mg
30136309 250 mg
30004179 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134323 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD965260500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 23255-15-4
Molekylformel C8H15NO5S
Molekylvikt (g/mol) 237.27
Synonym N-Acetyl-3-[(2,3-dihydroxypropyl)thio]-alanine
IUPAC-namn N-acetyl-S-(2,3-dihydroxypropyl)cysteine
LEDER CC(=O)NC(CSCC(O)CO)C(=O)O
Specifikationer
Kemiskt namn eller material DHPMA
InChI formel InChI=1S/C8H15NO5S/c1-5(11)9-7(8(13)14)4-15-3-6(12)2-10/h6-7,10,12H,2-4H2,1H3,(H,9,11)(H,13,14)
Kvantitet 500 mg
Formel vikt 237.0671