Läs mer
Di(2-(4-(dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl))-2-propoxyethyl Propionate, TRC
Produktkod. 30356278
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30356278 100 mg
30350413 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30356278 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD417110100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1798840-31-9
Molekylformel C40H42N6O3S2
Molekylvikt (g/mol) 718.9299
Synonym 1-Piperazineacetic acid, 4-dibenzo[b,f][1,4]thiazepin-11-yl-, 2-[2-(4-dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethyl ester, 2-[2-[4-(Dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]ethoxy]ethyl 2-[4-(dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl]acetate, Quetiapine Fumarate Imp. C (EP)
IUPAC-namn 2-[2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)ethoxy]ethyl 2-(4-benzo[b][1,4]benzothiazepin-6-ylpiperazin-1-yl)acetate
LEDER O=C(CN1CCN(CC1)C2=Nc3ccccc3Sc4ccccc24)OCCOCCN5CCN(CC5)C6=Nc7ccccc7Sc8ccccc68
Specifikationer
Kemiskt namn eller material Di(2-(4-(dibenzo[b,f][1,4]thiazepin-11-yl)piperazin-1-yl))-2-propoxyethyl Propionate
Kvantitet 100 mg
InChI formel InChI=1S/C40H42N6O3S2/c47-38(29-44-19-23-46(24-20-44)40-31-10-2-6-14-35(31)51-37-16-8-4-12-33(37)42-40)49-28-27-48-26-25-43-17-21-45(22-18-43)39-30-9-1-5-13-34(30)50-36-15-7-3-11-32(36)41-39/h1-16H,17-29H2
Formel vikt 718.276