Learn More
Di-2-pyridyl thionocarbonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.
Solubility
Soluble in water 9280 g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Di-2-pyridyl thionocarbonate is used in the synthesis of 10-des(carbamoyloxy)-10-isothiocyanatoporfiromycin. It is also used as a substitute of thiophosgene, in the preparation of isothiocyanate.
Solubility
Soluble in water 9280 g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 96°C to 99°C |
Quantity | 1 g |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in water 9280g/L). |
Formula Weight | 232.26 |
Percent Purity | 98% |
Chemical Name or Material | Di-2-pyridyl thionocarbonate |