Läs mer
MedChemExpress Di-8-ANEPPS
Specifikationer
Specifikationer
| Innehåll och lagring | -20°C, protect from light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Kvantitet | 1 mg |
| Frakt skick | Room temperature |
| Löslighet | DMSO : 1 mg/mL (1.69 mM; ultrasonic and warming and heat to 60°C) |
| Produkttyp | Dye Reagents |
| Molekylvikt (g/mol) | 592.87 g/mol |
| CAS | 157134-53-7 |