Läs mer
Di(Alpha-tocopherol) Phosphate, TRC
Produktkod. 30452213
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30452213 10 mg
30452852 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30452213 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT52619010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 311313-37-8
Molekylformel C58H99O6P
Molekylvikt (g/mol) 923.38
Synonym (2R,2'R)-3,4-Dihydro-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol 6,6'-(Hydrogen Phosphate)
IUPAC-namn bis[(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] hydrogen phosphate
LEDER CC1=C(C)C(OP(OC2=C(C)C(CC[C@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)O3)=C3C(C)=C2C)(O)=O)=C(C)C4=C1O[C@@](CCC[C@H](C)CCC[C@H](C)CCCC(C)C)(C)CC4
Specifikationer
Kemiskt namn eller material Di(alpha-tocopherol) Phosphate
Kvantitet 10 mg
InChI formel InChI=1S/C58H99O6P/c1-39(2)23-17-25-41(5)27-19-29-43(7)31-21-35-57(15)37-33-51-49(13)53(45(9)47(11)55(51)61-57)63-65(59,60)64-54-46(10)48(12)56-52(50(54)14)34-38-58(16,62-56)36-22-32-44(8)30-20-28-42(6)26-18-24-40(3)4/h39-44H,17-38H2,1-16H3,(H,59,60)/t41-,42-,43-,44-,57-,58-/m1/s1
Formel vikt 922.7179