Learn More
di-Ammonium Oxalate Monohydrate, MilliporeSigma™
Product Code. 15814050
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15814050 1 kg Plastic Bottle 1kg
15804050 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15814050 Supplier Supelco Supplier No. 1011921000
 Restricted Item
Beräknad leverans: 19-11-2026
For analysis EMSURE™ ACS, ISO, Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.111
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
Synonym ammonium oxalate monohydrate, ammonium oxalate monohydrate, acs, ethanedioic acid, diammonium salt, monohydrate, diazanium oxalate hydrate, acmc-1aw90, ksc493e5j, diammonium hydrate oxalate, oxalic acid ammonium salt monohydrate
PubChem CID 516808
IUPAC Name diazanium;oxalate;hydrate
SMILES C(=O)(C(=O)[O-])[O-].[NH4+].[NH4+].O
Specifications
Physical Form Solid
Quantity 1 kg
Solubility Information Soluble in water: 45g/L at 20°C
Formula Weight 142.11
Percent Purity 99.5% to 101.0%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Density 1.50g/cm3 at 20°C
Chemical Name or Material di-Ammonium oxalate monohydrate