missing translation for 'onlineSavingsMsg'
Learn More

Di-beta, beta'-kloroetylfosforsyra-d8, TRC

Product Code. 30350005
Change view
Click to view available options
Kvantitet:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30350005 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30350005 Supplier Toronto Research Chemicals Supplier No. TRCC3667921MG
 Begränsad artikel
Beräknad leverans: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Högrena organiska molekyler och analytiska standarder, strategiskt levererade världen över för att möjliggöra innovation och kommersiell framgång.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Di-beta,beta'-Chloroethylphosphoric Acid-d8
CAS 1477495-02-5
Anteckningar om renhetsgrad HPLC
Molekylformel C4HD8Cl2O4P
Kvantitet 1 mg
Synonym Bis(2-chloroethyl) Hydrogen Phosphate-d8, Bis(2-chloroethyl) Phosphate-d8, Bis(chloroethyl) Phosphate-d8, Bis(β-chloroethyl) Phosphate-d8, Bis(β-chloroethyl) Phosphoric Acid-d8, Di-2-Chloroethyl Phosphate-d8
InChI formel InChI=1S/C4H9Cl2O4P/c5-1-3-9-11(7,8)10-4-2-6/h1-4H2,(H,7,8)/i1D2,2D2,3D2,4D2
LEDER O=P(OC([2H])([2H])C([2H])([2H])Cl)(OC([2H])([2H])C([2H])([2H])Cl)O
IUPAC-namn bis(2-klor-1,1,2,2-tetradeutioetyl) vätefosfat
Molekylvikt (g/mol) 231.04
Formel vikt 230.0118
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.