Learn More
Di-n-butyl carbonate, Thermo Scientific Chemicals
407.63 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Density | 0.9200 g/mL |
Quantity | 5 g |
Specific Gravity | 0.92 |
Formula Weight | 174.24 |
Chemical Name or Material | Di-n-butyl carbonate |