Learn More
Di-n-butylmagnesium, 0.5M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
Catalog Number p-3212647
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
1191-47-5, 142-82-5, 97-93-8
C8H18Mg
138.54
MFCD00015225
KJJBSBKRXUVBMX-UHFFFAOYSA-N
dibutylmagnesium, magnesium, dibutyl, di-n-butylmagnesium, magnesium,butyl 1-methylpropyl, dibutyl magnesium, di-n-butyl magnesium, di-n-butylmagnesium solution, odhfjiddbsdwnu-uhfffaoysa-n, di-n-butylmagnesium, 0,5m in heptane, di-n-butylmagnesium solution, 1.0 m in heptane
70929
magnesium;butane
[Mg++].CCC[CH2-].CCC[CH2-]
1191-47-5, 142-82-5, 97-93-8
C8H18Mg
138.54
MFCD00015225
KJJBSBKRXUVBMX-UHFFFAOYSA-N
dibutylmagnesium, magnesium, dibutyl, di-n-butylmagnesium, magnesium,butyl 1-methylpropyl, dibutyl magnesium, di-n-butyl magnesium, di-n-butylmagnesium solution, odhfjiddbsdwnu-uhfffaoysa-n, di-n-butylmagnesium, 0,5m in heptane, di-n-butylmagnesium solution, 1.0 m in heptane
70929
magnesium;butane
[Mg++].CCC[CH2-].CCC[CH2-]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point -71.0°C
Density 0.7300g/mL
Flash Point −4°C
Packaging AcroSeal™ Glass bottle
Linear Formula (C4H9)2Mg
Quantity 100 mL
Specific Gravity 0.73
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Viscosity 0.9 mPa.s (22°C)
Formula Weight 138.54
Chemical Name or Material Di-n-butylmagnesium
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Di-n-butylmagnesium, 0.5M solution in heptane
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P273-Avoid release to the environment.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : n-Heptane, Triethylaluminum, Magnesium dibutyl-